Would you like to log in again?
PicoProbe™ Fructose-1,6-Bisphosphate Assay Kit (F), BioVision
Supplier: BioVision
AB284537-100TEST
76416-466EA 771.24 USD InStock
76416-466
PicoProbe™ Fructose-1,6-Bisphosphate Assay Kit (F), BioVision
Assays Enzyme Assays
To measure F1,6BP in a variety of sample types.
  • Simple and sensitive
  • Quick and easy way to measure Fructose-1,6-Bisphosphate in a variety of sample types
  • Easy to perform
Fructose-1,6-Bisphosphate (F1,6BP) is a fructose sugar and a key component of the glycolysis metabolic pathway. It is produced by the phosphorylation of Fructose 6-Phosphate (F6P) by phosphofructokinase (PFK). Additionally, F1,6BP is an allosteric activator of enzymes including PFK and pyruvate kinase. Furthermore, F1,6BP stimulates the Pentose Phosphate Pathway that produces glutathione and is associated with anti-convulsing effects in animal models. Recent studies have shown that F1,6BP can be taken up by various cell types thereby providing ATP in glycolysis without consuming ATP and preserving the organs such as heart, liver, kidney under ischemic conditions. BioVision’s PicoProbe™ Fructose-1,6-Bisphosphate Assay Kit provides a quick and easy way to measure F1,6BP in a variety of sample types. In this Assay, F1,6BP is hydrolyzed by Fructose-1,6-Bisphosphatase (FBP) into inorganic phosphate and F6P. F6P then through several intermediate reactions reacts with the PicoProbe™ to generate a stable fluorophore measured at Ex/Em = 535/587 nm. The assay is simple, easy to perform, sensitive and can detect F1,6BP less than 0.5 µM in a variety of samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty