Would you like to log in again?
Iodide Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB284538-100TEST
76416-468EA 742.92 USD InStock
76416-468
Iodide Colorimetric Assay Kit, BioVision
Assays Cellular Assays
To determine Iodide concentration
  • Convenient
  • Sensitive
  • Non-radioactive
  • Simple
  • Can detect as low as 0.2 μM Iodide under the assay conditions
Iodide (I−) is a halide anion and a necessary and limiting substrate for thyroid hormone synthesis. This essential element enables the thyroid gland to produce thyroid hormones, thyroxine (T4) and triiodothyronine (T3). Additionally, Iodide is important in many essential biological processes such as basal metabolism, temperature regulation, intellectual development for children, muscular development, normal heart function, growth of skeleton etc. Food is the main source of daily supply of iodide in humans. Iodide can be found in grains, fish, dairy products, fruits, iodized salts etc. Iodine deficiency affects around two billion people worldwide. Iodide deficiency can cause diseases such as goiter, depression, fatigue, hypothyroidism, intellectual disability, delayed sexual development etc. However, excess intake of Iodide is also harmful and can inhibit the release of thyroid hormones and cause inflammation of the thyroid gland and even thyroid cancer. BioVision’s Iodide Colorimetric Assay Kit provides a quick, convenient, non-radioactive method to determine Iodide concentration in various sample types including foods, liquid and biological samples. The assay relies on the Iodide ion as a specific catalyst to convert a yellow colored substrate to a colorless product. The reduction in absorbance signal at 330 nm is directly proportional to the Iodide concentration in the samples. Most common ion do not interfere with the assay. The kit is simple, rapid, sensitive and can detect as low as 0.2 μM Iodide under the assay conditions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty