Would you like to log in again?
Laccase Activity Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB284539-100TEST
76416-470EA 877.36 USD InStock
76416-470
Laccase Activity Assay Kit (Colorimetric), BioVision
Assays Enzyme Assays
To measure Laccase activity in various samples
  • Rapid and easy-to-use
  • High-throughput adaptable
  • Can measure Laccase activity as low as 20 µU in samples.
Laccases (EC 1.10.3.2), also known as p-benzenediol: oxygen oxidoreductases are a class of copper-containing enzymes found in bacteria, insects, plants and fungi. They catalyze the oxidation of aromatic compounds using molecular oxygen as the terminal electron acceptor and their molecular mass ranges from 60-100 kDa. Laccases play a key role in lignin synthesis and degradation and are utilized in several biotechnological applications such as degradation of dyes, chemical wastes, xenobiotics and other phenolic compounds. Additionally, Laccases are used in food industry for producing cost-effective, healthy foods. Furthermore, they are also being investigated in the construction of Laccase biosensors because of their high redox potential. BioVision’s Laccase Activity Assay Kit is a simple, one step, plate based assay to measure Laccase activity in various samples. In this assay, samples containing Laccase will oxidize the substrate to generate a colored product with a strong absorbance at 420 nm. The kit is rapid, easy to use and high-throughput adaptable. It can measure Laccase activity as low as 20 µU in samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty