Would you like to log in again?
Alpha-Mannosidase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB282917-100TEST
76416-476EA 997.67 USD InStock
76416-476
Alpha-Mannosidase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
To monitor total α-Mannosidase activity
  • Can detect as low as 1.0 µU of alpha mannosidase activity
  • Facile, rapid way to monitor total Alpha-Mannosidase in various biological samples
  • The substrate is specific to α-Man and can differentiate from β-mannosidase
Alpha-Mannosidase (α-Man; EC 3.2.1.24) is a glycoside hydrolase involved in the cleavage of the alpha-mannose. α-Man can be applied in studies that determine the effects of mannose on specific molecules, such as recombinant proteins that are used for vaccine development. Lysosomal α-Man is essential for catabolizing the Asn-linked glycans of glycoproteins and is actively involved in maintaining the cellular homeostasis. α-Man deficiency results in Alpha-Mannosidosis, an acquired lysosomal storage disorder in which α-Man fails to break down mannose-containing oligosaccharides. These oligosaccharides, therefore, accumulate in lysosomes thereby leading to cellular malfunction and apoptosis. BioVision’s Alpha-Mannosidase Activity Assay Kit provides a facile, rapid way to monitor total α-Man activity in various biological samples. In this kit, α-Man cleaves a synthetic specific substrate and releases a fluorophore, which can be easily quantified (Ex/Em = 360/445 nm). The substrate is specific to α-Man and can differentiate from β-mannosidase. The assay is simple, sensitive and can detect as low as 1.0 µU of alpha mannosidase activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty