Would you like to log in again?
Ionizer Blower, Sartorius
Supplier: Sartorius
YIB01-0UR
76413-010EA 2440.98 USD InStock
76413-010
Ionizer Blower, Sartorius
Ionizers
Ionizing blower for neutralize static electricity within a few seconds, by 'shooting' ions at the material to be weighed. This eliminates the need for complicated, expensive, and often ineffective, increasing of the humidity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty