Would you like to log in again?
PIG® White Meltblown Pads , NEW PIG
Supplier: New Pig
PIG®
MAT862
89208-794CS 127.35 USD InStock
89208-794
PIG® White Meltblown Pads , NEW PIG
Spill Control Materials
Soak up oil and save money with these economical meltblown mat pads.
  • Fast-wicking fibers pull oil from the surface of water
  • Water-repellent meltblown polypropylene is highly absorbent for cleaning up oil and oil-based fluids
  • Floats even when it's soaked; ideal for spill response on bodies of water
  • Bright white color is easy to find in the event of a spill
The mat stays together, even when fully saturated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty