Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crimp Top Vials and Caps, 2 ml, Agilent Technologies

Supplier: Agilent Technologies
5190-9064 8010-0196 5183-4494 5183-4491 5181-3400 5181-3375 9301-1419 5183-4497 5181-3376 5182-0543 5190-4035 5190-2241 5190-2282 5190-4033 5183-4496 5181-8801 5190-9064 5190-4036 5183-4492 5183-4492 5190-9558 5190-6113 8010-0195 5067-0214 5190-6113 5183-4497 5183-4491 5190-2282 5190-4032 5181-3375 5190-6123 5190-4036 5190-2241 5190-9065 5183-4495 5181-3400 5190-4033 5181-8801 5190-4032 5182-0543 5183-4494 5190-6117 5190-2283 8010-0196 5183-4495 5190-9065 5183-4493 5185-5852 5190-2283 5190-6116 5190-9558 5190-6182 5181-3376 5183-4493 5185-5946 5190-6116 5190-4035 8010-0195 5190-4046 5183-4496 9301-1419 5190-4046
AG5183-4494PK 50.45 USD InStock
AG5183-4494 AG5190-4046 AG5183-4493 AG5183-4492 AG5183-4491 AG5183-4497 AG5183-4496 AG5183-4495 CA76514-744 CA8010-0196 CA8010-0195 AG5181-8801 103583-918 CA5190-4032 103584-492 CA5190-4036 CA76515-146 101409-604 CA5190-4033 103582-808 AG5190-4035 AG5190-4036 AG5190-4033 103582-800 CAAG5190-6113 CAAG5190-6116 CAAG5190-9064 AG5181-3400 CAAG5190-9065 CA5190-4046 AG5190-4032 AG5190-2241 AG5182-0543 CAAG5190-2241 AG8010-0196 AG8010-0195 AG5190-9558 CA5183-4494 CA5181-8801 103584-502 CA5183-4497 CA76514-882 AG5181-3375 AG5181-3376 CA76508-098 AG5190-2283 AG5190-2282 AG5190-6113 CA76514-758 AG5190-6116 CA76514-876 CA76514-878 CAAG5190-4035 CA76514-750 CA76514-874 CA76514-880 CA5190-2282 CA5190-2283 AG9301-1419 AG5190-9064 CA5190-9558 AG5190-9065
Crimp Top Vials and Caps, 2 ml, Agilent Technologies
Vials
These 2 ml crimp top vials and matching crimp caps are engineered to protect against sample loss.

  • Multiple crimp cap colors and septa materials to fit with a wide range of 2 ml crimp vials
  • All feature wide opening crimp caps providing a larger target area for autosampler
  • Guaranteed to work effectively with 10 autosampler brands

Agilent 2 ml crimp top vials are constructed from Type 1 borosilicate glass, with low metal content, to protect your sample from destabilizing or leaching.

The associated crimp caps come with a choice of either PTFE/silicone, PTFE only, natural red rubber, butyl, or PTFE/butyl septum materials.

The 2 ml crimp top vials and 11 mm crimp caps are the perfect fit for HPLC and GC instrument platforms across a wide variety of applications, including pharmaceutical, environmental, energy and fuels, forensics, materials science, biopharmaceutical, proteomics, and metabolomics. The crimp style vial is sometimes referred to as the GC vial.

Certifications: Compliant with U.S. and European Pharmacopeia requirements.

Packaging: Packed in a clean environment with packaging specially designed to reduce breakage and prevent contamination.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty