Would you like to log in again?
Eclipse 5300 Automated Dissolution Samplers, Distek
Supplier: Distek
2022-5300-8-V 2022-5300-V 2022-5300-VF 2022-5300-8-VF
76419-878EA 40823.33 USD InStock
76419-878 76419-874 76419-876 76419-880
Eclipse 5300 Automated Dissolution Samplers, Distek
Autosamplers
The Distek Eclipse 5300 automated dissolution sampler offers rapid, accurate sampling using precision syringe pumps. The elimination of valves and rotary pumps, along with the optimized sample pathway with a minimal 4.5 ml internal volume, increases reliability while reducing common sources of sample carryover.
  • Rapid and accurate sampling
  • No valves or rotary pumps
  • Cost-effective dual bath solution
  • Media recycling and replacement
  • Color touch screen display
  • Optional filter changer
Adding a lower-cost ‘child’ sampler is a cost-effective way to sample from two baths. Each of the baths may run the same method, or two different, independent ones. The integrated optional filter changer enables the use of most common 25 mm syringe filters instead of requiring custom filter plates and additional bench space.
Certifications: ETL, CE.
Ordering information: Includes the following accessories; test tube and vial racks, 500 ml and 900 ml sampling probes, 10 and 45 µm filters. Includes a 1 year comprehensive warranty. Does not include; installation or qualification services.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty