Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Coviblock COVID-19 IgG-IgM Antibody Test, Clarity

Supplier: Clarity Diagnostics
CD-COV19 CDCOV19IGGCTL
76423-340BX 186.41 USD InStock
76423-340 76423-342
Coviblock COVID-19 IgG-IgM Antibody Test, Clarity
Clinical Diagnostic Supplies Clinical Reagents and Kits
  • Sensitivity 93.3%
  • Specificity 98.8%
  • Use whole blood, plasma or serum
  • Fast and easy (with positive results as fast as 10 minutes)
  • Room temperature storage

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty