Would you like to log in again?
Bel-Art Sterileware® Double Bagged Sterile Scoop Sampling Systems, SP Scienceware
Supplier: Bel-Art Products, a part of SP
Sterileware®
H36924-1010 H36922-1010
76423-420CS 85.42 USD InStock
76423-420 76423-418
Bel-Art Sterileware® Double Bagged Sterile Scoop Sampling Systems, SP Scienceware
Scoops
Bagged twice for extra protection from contamination. Ten individually packaged scoops are sealed in an outer bag to create multiples layers of protection against contaminants and debris.
  • Snap-on lid allows scoop to be used as a container for transport; lock-tab can be bent under for a tighter seal
  • Sampling data can be marked on the lid with wax pencil, pre-marked label or labeling tape to facilitate tracking
  • Scoops remain upright when placed on a flat surface; can be used as weighing boats
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10⁻⁶
  • Lot stamped for accurate lot tracing, and shipped with a certificate of processing for sterilization
The long handle increases the distance between the hand and sample to reduce contamination potential. Double bagging reduces contamination in multi-zone clean rooms while the included lid eliminates the cost of separate sterile bags or containers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty