Would you like to log in again?
SP Bel-Art® Sterileware® Powder Probe Samplers, Bel-Art Products, a part of SP
Supplier: Bel-Art
Sterileware®
F36975-1000 F36975-0600
76402-898PK 193.65 USD InStock
76402-898 76402-900
SP Bel-Art® Sterileware® Powder Probe Samplers, Bel-Art Products, a part of SP
Samplers Solid Samplers
Ideal for taking a cross sectional sample through a bulk powder, this probe is available in two convenient lengths.
  • End tip contains free-flowing powders within the probe
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) probe is heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Used by pharmaceutical, food, cosmetic and chemical companies.
Ordering information: Sterilized products are not returnable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty