Acrolein and Acrylonitrile Standards for US EPA Method 603, VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0715-1ML BDH0753-1ML
BDH0715-1MLEA 71.81 USD InStock
BDH0715-1ML BDH0753-1ML
Acrolein and Acrylonitrile Standards for US EPA Method 603, VWR Chemicals BDH®
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 603 is a general analytical method for the separation of acrolein and acrylonitrile in municipal and industrial wastewater by GC/FID.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty