Would you like to log in again?
Aroclor 1232 for US EPA Methods 505, 508, 608, 8082, CLP Series Methods and ASTM Method D6160
Supplier: Avantor
BDH0788-1ML
BDH0788-1MLEA 59.06 USD InStock
BDH0788-1ML
Aroclor 1232 for US EPA Methods 505, 508, 608, 8082, CLP Series Methods and ASTM Method D6160
Standards Organic Standards
Semivolatiles standard.
US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty