Would you like to log in again?
Purgeable Gases Mix B for US EPA Methods 502.1, 502.2, 601, 624, 8021, 8260 and CLP Series Methods
Supplier: Avantor
VWR Chemicals BDH®
BDH0721-1ML
BDH0721-1MLEA 34.48 USD InStock
BDH0721-1ML
Purgeable Gases Mix B for US EPA Methods 502.1, 502.2, 601, 624, 8021, 8260 and CLP Series Methods
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 502.1 is a general analytical method for the monitoring of halogenated, portable VOCs in drinking water and raw water sources by GC with a Hall Detector.
US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 601 is a general analytical method for the monitoring of halogenated VOCs in municipal and industrial wastewater by GC with a Hall Detector.
US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.
US EPA Method 8021 is a general analytical method for the monitoring of VOCs by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty