Would you like to log in again?
Trihalomethanes Volatiles Mix for US EPA Method 502.2
Supplier: Avantor
VWR Chemicals BDH®
BDH0804-1ML
BDH0804-1MLEA 33.28 USD InStock
BDH0804-1ML
Trihalomethanes Volatiles Mix for US EPA Method 502.2
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty