Would you like to log in again?
KIMBLE® NMR Tube Rack, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
897080-0005
KT897080-0005EA 95.88 USD InStock
KT897080-0005
KIMBLE® NMR Tube Rack, DWK Life Sciences
Racks Tube Racks
PP NMR tube rack.
  • Accommodates 72 3 or 5 mm tubes
Rack with carrying handles features an upper deck with holes and a lower deck support for tube ends.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty