Would you like to log in again?
Ruhof Viral Transport Media, Clarity Diagnostics
Supplier: Clarity Diagnostics
345TK-M
76436-480CS 3054.13 USD InStock
76436-480
Ruhof Viral Transport Media, Clarity Diagnostics
Media for Microbiology Prepared Media for Microbiology
Active viral transport medium offered by Ruhof corporation is for the preservation and transportation of viruses, such as coronavirus (Covid-19).
  • 18 month shelf life
  • Room temperature stable
  • Available in three ml
  • Provides a pH balanced environment
  • A phenol red pH indicator provides a visual gauge of the medium integrity throughout the storage and life of the product
  • Includes antibiotics to inhibit bacteria and fungal flora
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty