Would you like to log in again?
Disposable Face Shield, Cole Parmer
Supplier: Cole Parmer
78900-46
76436-438CS 593.66 USD InStock
76436-438
Disposable Face Shield, Cole Parmer
Face Shields
Wrap-around design provides full facial protection.
  • Foam padded headband provides extra comfort
  • Clear PET delivers maximum visibility; remove protective film before use
  • Suitable to wear over eyeglasses
The disposable face shield protects eyes, mouth, and nose from droplets, sprays, aerosols, splatters, and flying debris. Transparent PET shield is lightweight, breathable, and comfortable to wear. The 1.5" elastic nylon strap and foam-padded headband allow for comfortable, extended wear.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty