Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boric acid 20 g/L, Snip & Pour™

Supplier: ENVIRONMENTAL EXPRESS
Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

C7120
76436-852EA 208.89 USD InStock
76436-852
Boric acid 20 g/L, Snip & Pour™
ortho-Boric acid
Premeasured Snip & Pour reagent tubes for ammonia and cyanide distillations, deliver the precise volume of reagent to each sample.

  • Easy-to-use, just snip and pour into receiver tube
  • Increase accuracy by eliminating the need to measure and pipette
  • Packaged in convenient PFA tubes


Formula: B(OH)₃
MW: 61.83 g/mol
MDL Number: MFCD00011337
CAS Number: 10043-35-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty