Would you like to log in again?
Legacy Everwipe Disinfectant Wipes, Essendant
Supplier: Essendant
Legacy
LEY10100
76442-800CT 236.42 USD InStock
76442-800
Legacy Everwipe Disinfectant Wipes, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
The Everwipe Disinfectant Wipe is an EPA registered, hospital-grade disinfectant that quickly and effectively disinfects hard, non-pourous surfaces. Kills bacteria, viruses, and fungi.
  • Cleans, disinfects and deodorizes
  • Alchohol free, bleach free
  • Quat based formulation
  • Application: General purpose
Applicable material: Hard, non-porous, inanimate environmental surfaces; floors; walls; metal surfaces; stainless steel; glazed porcelain; glazed ceramic tile; plastic; bathroom shower stalls.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty