Would you like to log in again?
Volatile ISTD Mix for US EPA Methods 624 and 8240, SPEX CertiPrep
Supplier: SPEX CertiPrep
ECS-A-010 ECS-B-010
SXECS-A-010EA 61.4 USD InStock
SXECS-A-010 SXECS-B-010
Volatile ISTD Mix for US EPA Methods 624 and 8240, SPEX CertiPrep
Standards Environmental Analysis Standards
ECS Standards for GC/MS
All ampulized standards are supplied with a pre-labeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty