Would you like to log in again?
ISTD Mix for US EPA Method 8260B, SPEX CertiPrep
Supplier: SPEX CertiPrep
ECS-A-051 ECS-B-051
SXECS-B-051EA 137.2 USD InStock
SXECS-B-051 SXECS-A-051
ISTD Mix for US EPA Method 8260B, SPEX CertiPrep
Standards Environmental Analysis Standards
ECS Standards for GC/MS
All ampulized standards are supplied with a pre-labeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date.
1,4-Dichlorobenzene-d43855-82-12000 µg/ml
Chlorobenzene-d53114-55-42000 µg/ml
Fluorobenzene462-06-62000 µg/ml
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty