Would you like to log in again?
Organic Wine Standard Set 1, SPEX CertiPrep
Supplier: SPEX CertiPrep
WINE-1
SXWINE-1EA 163.95 USD InStock
SXWINE-1
Organic Wine Standard Set 1, SPEX CertiPrep
Standards Food Standards
To help regulate the spoilage of wine, the wine industry tests for several compounds that are known to cause this. For this industry, SPEX CertiPrep offers Certified Reference Materials (CRMs) for wine that are designed for GC, GC/MS, HPLC and LC/MS analysis. These standards are manufactured from the highest purity starting materials and the highest grade of solvents available in order to guarantee superior quality. Each standard is supplied with a detailed comprehensive Certificate of Analysis.
  • Consumer Safety Standards
  • Analytical Standards for Wine
Wine is a complex combination of water, alcohol and other substances such as organic acids, phenols, sugars, carbon dioxide, and sulfur dioxide. The chemical interactions of these compounds, combined with the aging process, determine the quality of the color, taste and aroma of the wine. However, during this process, the chemical interactions can also play key roles in spoiling the wine. For instance, cork taint transpires when naturally occurring airborne fungi are in the presence of chlorophenol compounds to form 2,4,6-trichloranisole (TCA) in the wine. When this occurs, the wine’s aroma is reduced significantly and is replaced by an undesirable smell and taste.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty