Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrofluoric acid 48.50 - 49.50 %, CMOS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = R

9585-69 BAKR9585-69
JT9585-69EA 3.73 USD InStock
JT9585-69 CAJT9585-69
Hydrofluoric acid 48.50 - 49.50 %, CMOS, J.T.Baker®
Hydrofluoric acid


Formula: HF
MW: 20.01 g/mol
Melting Pt: –36 °C
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II
Order Now
Specification Test Results
For Microelectronic Use
Assay (HF) 48.50 - 49.50 %
Fluosilicic Acid (H₂SiF₆) ≤50 ppm
Chloride (Cl) ≤5000 ppb
Nitrate (NO₃) ≤3000 ppb
Phosphate (PO₄) ≤1000 ppb
Sulfate (SO₄) ≤5000 ppb
Trace Impurities - Arsenic (As) ≤10.0 ppb
Trace Impurities - Antimony (Sb) ≤10.0 ppb
Trace Impurities - Aluminum (Al) ≤10.0 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤10.0 ppb
Trace Impurities - Calcium (Ca) ≤10.0 ppb
Trace Impurities - Chromium (Cr) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Iron (Fe) ≤10.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤10.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤10.0 ppb
Trace Impurities - Sodium (Na) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤10.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤10.0 ppb

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty