Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 96%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

MACR2874-R 2874-R MACR2874-R
JT2874-REA 3.87 USD InStock
JT2874-R CAMK2874-R MK2874-R
Sulfuric acid 96%, AR® ACS, Macron Fine Chemicals™
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (H₂SO₄) 95.0 - 98.0 %
ACS - Color (APHA) ≤8
Identification A Passes Test
Identification B Passes Test
ACS - Substances Reducing Permanganate (as SO₂) Passes Test
ACS - Residue after Ignition ≤4 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤0.5 ppm
ACS - Ammonium (NH₄) ≤1 ppm
ACS - Chloride (Cl) ≤0.1 ppm
ACS - Nitrate (NO₃) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤0.200 ppm
Trace Impurities - Arsenic (As) ≤0.004 ppm
Trace Impurities - Boron (B) ≤0.010 ppm
Trace Impurities - Chromium (Cr) ≤0.200 ppm
Trace Impurities - Copper (Cu) ≤0.10 ppm
Trace Impurities - Gold (Au) ≤0.300 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm
Trace Impurities - Lead (Pb) ≤0.300 ppm
Trace Impurities - Magnesium (Mg) ≤0.20 ppm
Trace Impurities - Manganese (Mn) ≤0.200 ppm
Trace Impurities - Nickel (Ni) ≤0.1 ppm
Trace Impurities - Potassium (K) ≤0.300 ppm
Trace Impurities - Tin (Sn) ≤0.200 ppm
Trace Impurities - Titanium (Ti) ≤0.300 ppm
Trace Impurities - Zinc (Zn) ≤0.200 ppm
Trace Impurities - Mercury (Hg) ≤5.0 ppb

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty