Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ferric pyrophosphate soluble, pearls, purified, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Iron(III) pyrophosphateIron (III) pyrophosphate

Management of Change (MOC) category = R

MACR5528-25 MACR5528-25
JT5528-25CS 10994.7 USD InStock
JT5528-25 MK5528-25
Ferric pyrophosphate soluble, pearls, purified, Macron Fine Chemicals™
Ferric pyrophosphate


Formula: Fe₄O₂₁P₆
MW: 745.2100 g/mol
MDL Number: MFCD00016091
CAS Number: 10058-44-3
Order Now
Specification Test Results
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (Fe) 10.5 - 12.5 %
Solubility Passes Test
Identification A Passes Test
Identification B Passes Test
Ammonium Salts Passes Test
Arsenic (As) ≤0.0008 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty