Would you like to log in again?
Alera® True HEPA Air Purifier Replacement Filter, Essendant
Supplier: Essendant
Alera®
ALEAPFILTER
76436-630PK 56.7 USD InStock
76436-630
Alera® True HEPA Air Purifier Replacement Filter, Essendant
Filters Gas Filters
This True HEPA filter removes up to 99.97% of microscopic airborne particles.
  • Designed to fit Alera® 4-speed HEPA Air Purifier.
  • Replaceable HEPA filter lasts 720 hours
  • Easy to install
Designed to fit the Alera® 4-speed HEPA Air Purifier. For optimal results, replace filter every 720 hours.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty