Would you like to log in again?
High Performance FR Women's Dual Hazard Lightweight Pants, Navy, Lakeland Industries
Supplier: Lakeland Industries
IP8WWUT13-8-30 IP8WWUT13-20-34 IP8WWUT13-18-28 IP8WWUT13-6-34 IP8WWUT13-4-32 IP8WWUT13-16-32 IP8WWUT13-16-28 IP8WWUT13-6-30 IP8WWUT13-12-34 IP8WWUT13-8-32 IP8WWUT13-0-30 IP8WWUT13-6-32 IP8WWUT13-4-30 IP8WWUT13-14-28 IP8WWUT13-20-32 IP8WWUT13-10-34 IP8WWUT13-14-30 IP8WWUT13-18-34 IP8WWUT13-14-32 IP8WWUT13-2-32 IP8WWUT13-12-32 IP8WWUT13-12-30 IP8WWUT13-20-28 IP8WWUT13-16-30 IP8WWUT13-10-32 IP8WWUT13-10-30 IP8WWUT13-20-30 IP8WWUT13-6-28 IP8WWUT13-0-32 IP8WWUT13-10-28 IP8WWUT13-16-34 IP8WWUT13-18-32 IP8WWUT13-8-34 IP8WWUT13-2-34 IP8WWUT13-0-34 IP8WWUT13-2-28 IP8WWUT13-14-34 IP8WWUT13-12-28 IP8WWUT13-4-28 IP8WWUT13-2-30 IP8WWUT13-8-28 IP8WWUT13-18-30 IP8WWUT13-4-34 IP8WWUT13-0-28
76438-278EA 139.21 USD InStock
76438-278 76438-356 76438-312 76438-334 76438-314 76438-336 76438-358 76438-338 76438-316 76438-318 76438-282 76438-360 76438-284 76438-362 76438-340 76438-286 76438-364 76438-342 76438-320 76438-288 76438-280 76438-308 76438-300 76438-344 76438-322 76438-346 76438-302 76438-324 76438-304 76438-326 76438-348 76438-306 76438-328 76438-292 76438-294 76438-350 76438-296 76438-352 76438-330 76438-298 76438-332 76438-310 76438-354 76438-290
High Performance FR Women's Dual Hazard Lightweight Pants, Navy, Lakeland Industries
Workwear
High Performance FR Lightweight Pants from Lakeland feature 8 oz. Westex® UltraSoft® fabric that has a unique basketweave blend and provides increased mobility and comfort, making them an excellent choice for arc flash and flash fire protection.
  • Wider waistband for increased comfort
  • Articulated knees offers better mobility
  • Cell phone pocket on right leg
  • Stealth zipper pocket on right leg
  • Crotch gusset for increased comfort
  • Extra material in heal to reduce wear
Lakeland FR Lightweight Pants are made with Westex® UltraSoft® and are NFPA 2112, UL certified, and meet NFPA 70E, CAT 2, making them an excellent choice for arc flash and flash fire protection. The fabric has a unique basketweave blend that provides increased mobility and comfort. Attention to design with features like an articulated knee, wider waistband, and a crotch gusset ensure all day comfort.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 9.8 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty