Would you like to log in again?
High Performance FR Women's Dual Hazard Lightweight Pant, Khaki, Lakeland Industries
Supplier: Lakeland Industries
IP8WWUT20-12-30 IP8WWUT20-4-28 IP8WWUT20-0-30 IP8WWUT20-12-32 IP8WWUT20-16-34 IP8WWUT20-12-34 IP8WWUT20-6-34 IP8WWUT20-2-32 IP8WWUT20-16-32 IP8WWUT20-6-32 IP8WWUT20-18-34 IP8WWUT20-18-32 IP8WWUT20-8-28 IP8WWUT20-18-30 IP8WWUT20-20-32 IP8WWUT20-14-30 IP8WWUT20-18-28 IP8WWUT20-4-30 IP8WWUT20-10-34 IP8WWUT20-4-32 IP8WWUT20-20-28 IP8WWUT20-8-34 IP8WWUT20-4-34 IP8WWUT20-0-32 IP8WWUT20-10-32 IP8WWUT20-14-28 IP8WWUT20-8-30 IP8WWUT20-8-32 IP8WWUT20-6-30 IP8WWUT20-20-34 IP8WWUT20-2-34 IP8WWUT20-16-28 IP8WWUT20-16-30 IP8WWUT20-14-34 IP8WWUT20-12-28 IP8WWUT20-10-28 IP8WWUT20-14-32 IP8WWUT20-6-28 IP8WWUT20-10-30 IP8WWUT20-20-30 IP8WWUT20-2-28 IP8WWUT20-2-30
76438-418EA 129.92 USD InStock
76438-418 76438-410 76438-432 76438-378 76438-434 76438-412 76438-414 76438-436 76438-438 76438-416 76438-380 76438-382 76438-384 76438-440 76438-386 76438-442 76438-420 76438-408 76438-366 76438-388 76438-400 76438-422 76438-444 76438-368 76438-402 76438-446 76438-424 76438-426 76438-404 76438-448 76438-406 76438-428 76438-392 76438-370 76438-394 76438-372 76438-374 76438-396 76438-430 76438-398 76438-376 76438-390
High Performance FR Women's Dual Hazard Lightweight Pant, Khaki, Lakeland Industries
Workwear
High Performance FR Lightweight Pants from Lakeland feature 8 oz. Westex® UltraSoft® fabric that has a unique basketweave blend and provides increased mobility and comfort, making them an excellent choice for arc flash and flash fire protection.
  • Wider waistband for increased comfort
  • Articulated knees offer better mobility
  • Cell phone pocket on right leg
  • Stealth zipper pocket on right leg
  • Crotch gusset for increased comfort
  • Extra material in heal to reduce wear
Lakeland FR Lightweight Pants are made with Westex® UltraSoft® and are NFPA 2112, UL certified, and meet NFPA 70E, CAT 2, making them an excellent choice for arc flash and flash fire protection. The fabric has a unique basketweave blend that provides increased mobility and comfort. Attention to design with features like an articulated knee, wider waistband, and a crotch gusset ensure all day comfort.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 9.8 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty