Would you like to log in again?
High Performance FR Short Sleeve Knit Crew, Khaki, Lakeland Industries
Supplier: Lakeland Industries
SSCAT20-MDT SSCAT20-4X SSCAT20-4XT SSCAT20-2XT SSCAT20-3XT SSCAT20-5X SSCAT20-XL SSCAT20-3X SSCAT20-MD SSCAT20-2X SSCAT20-XLT SSCAT20-XS SSCAT20-LGT SSCAT20-6X SSCAT20-SM SSCAT20-LG
76439-220EA 141.41 USD InStock
76439-220 76439-210 76439-232 76439-230 76439-228 76439-218 76439-226 76439-216 76439-238 76439-224 76439-214 76439-236 76439-222 76439-234 76439-212 76439-208
High Performance FR Short Sleeve Knit Crew, Khaki, Lakeland Industries
Workwear
Lightweight, inherently FR Short Sleeve Knit Crew with permanent moisture wicking properties make this an extremely comfortable FR/AR base layer garment.
  • Unparalleled CAT 2 protection at 16.4 cal/cm²
  • Lightweight 6.95 oz. inherently FR fabric
  • Excellent breathability
  • Permanent moisture wicking properties
  • Tag-free collar
  • Drop shoulder design
  • Left chest pocket
Base layers are the foundational key to the Lakeland FR/AR Layering System. To keep cool during hot summer days, and warm when it's frigid cold outside, wearers need to be dry. Base layer shirts have the optimal 60/40 blend of hydrophobic and hydrophilic fibers to constantly pull moisture away from the body, move it to the outer surface of the garment, and allow it to dry. Staying dry is the key to keeping wearers comfortable and better protected.
Certifications: NFPA 2112 - UL certified. ATPV: 16.4 cal/cm². CAT: 2.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty