Would you like to log in again?
Oxivir® TB Ready to Use Disinfetant, Cherry Almond Scent, Diversey, Essendant
Supplier: Essendant
DVO101104055
76452-012EA 299.46 USD InStock
76452-012
Oxivir® TB Ready to Use Disinfetant, Cherry Almond Scent, Diversey, Essendant
Cleaning Agents Disinfectants
Ready-to-use disinfectant cleaner based on proprietary hydrogen peroxide (AHP®) technology to deliver fast, effective cleaning performance.
  • One-step cleaner disinfectant powered by AHP® technology.
  • Effective against key pathogens,TB, MRSA, Norovirus and VRE.
  • Kills HBV, HCV and HIV in one minute, Tuberculocidal in five minutes and Fungicidal in ten minutes.
  • Compatible with most hard, non-porous surfaces.
  • 30 second soft-surface sanitizer
Disinfects in 60 seconds. Virucide, bactericide, tuberculocide, fungicide and non-food contact sanitizer. Kills MRSA and Norovirus. Meets bloodborne pathogen standards for decontaminating blood and body fluids. Colorless with a characteristic scent.
Application: Disinfectant/Cleaner.
Applicable Material: Hard, Non-Porous Surfaces.
Chemical Compound: Benzyl Alcohol; Hydrogen Peroxide.
Dirt Types: Irucide; Bactericide; Tuberculocide; Fungicide.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty