Would you like to log in again?
AS950 Portable Compact Samplers, Hach
Supplier: Hach
ASPCXXXC221XX ASP.CXXXC261XX ASPCXXXC261XX ASP.CXXXC121XX ASP.CXXXC161XX ASP.CXXXC221XX
76452-228EA 8041.95 USD InStock
76452-228 76452-226 76452-224 76452-230
AS950 Portable Compact Samplers, Hach
Autosamplers
The AS950 compact portable sampler is a reliable collection tool for many field applications.
  • An essential tool for viral detection in wastewater
  • Representative sampling is critical to a reliable COVID-19 community spread program
  • COVID-19 sampling of wastewater helps in detection (days before symptoms)
  • Varying sample sizes at user-specified time intervals
  • Simple setup with multiple power options
  • Standard memory and USB allow program uploads and data to be logged and transferred - no laptop needed!
  • Large full-color intuitive display for virtually error-free programming
Automatic sampling makes testing easy. The AS950 portable compact 2.5 gallon 12 V sampler simplifies water collection with a large, full-color display. The intuitive interface allows for single-screen programming, USB upload/download capabilities, and the ability to copy programs from one sampler to another, saving time. An error alert and status screen eliminate uncertainty and simplify troubleshooting, providing confidence in your sampling. Weighing only 28 pounds (12.7 kg), the AS950 portable sampler 12 V is a portable sampler capable of operation in remote settings on its battery.
Applications: Typically used in pre-treatment water sampling or for compliance in wastewater, the AS950 Portable Compact is also capable of monitoring SARS-CoV-2 in sewage. Composite sampling serves as a comparison and may indicate an early warning of an outbreak. Once collected, sewage is ready to be further analyzed for viral loading and specific RNA sequences of COVID-19.
Easiest and most intuitive operation. Most convenient data transfer and programming available. Confidence in your sampling process. Field convertible for compact or discrete sampling. Easy maintenance at low cost.
Certifications: UL and CE certified.
Accessories information: Battery charger (76452-216) sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty