Would you like to log in again?
NG-Test® CARBA 5 Rapid Kit NG Test For Detection, Hardy Diagnostics
Supplier: Hardy Diagnostics
CARBA5
76375-810EA 571.37 USD InStock
76375-810
NG-Test® CARBA 5 Rapid Kit NG Test For Detection, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
NG-Test® CARBA 5 is an in vitro rapid and visual multiplex immunochromatographic assay for the qualitative detection and differentiation of five common carbapenemases (KPC, OXA-48-like, VIM, IMP and NDM) from carbapenem non-susceptible pure bacterial colonies.
  • Detects the gene expression which can be crucial to aiding therapeutic decisions
CARBA 5 is the only rapid, multiplex, phenotypic test capable of detecting KPC, OXA-48-like, VIM, IMP, and NDM carbapenemases produced by Enterobacterales and P. aeruginosa.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty