Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride solution 0.9% USP for injection

Supplier: Fresenius Kabi
Synonyms: HaliteSea saltSaline

65219-470-30 63323-623-75 63323-623-61 63323-623-76 63323-623-53
76460-918CS 352.27 USD InStock
76460-918 76460-926 76460-924 76460-922 76460-920
Sodium chloride solution 0.9% USP for injection
Sodium chloride
  • Preservative Free
  • Bar Coded
  • AP Rated
  • Non-DEHP
  • Non-PVC


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty