Would you like to log in again?
First Aid Only™ NuStat Hemostatic Gauze Pad , Acme United
Supplier: First Aid Only
First Aid Only™
91196-002
76452-344EA 33.81 USD InStock
76452-344
First Aid Only™ NuStat Hemostatic Gauze Pad , Acme United
First Aid Supplies First Aid Kit Components
The NuStat Hemostatic gauze pad is designed to promote rapid blood coagulation in the event of a traumatic wound involving an arterial bleed. 
  • Promotes rapid blood coagulation
The 4×4" Hemostatic pad's patented and natural formula works independently of the body's normal clotting mechanisms to clot blood even where normal clotting is slow or impaired. Applying the pad and applying pressure is a safe, easy and very effective solution to traumatic bleeding situations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty