Would you like to log in again?
Monarch® DNA/RNA Protection Reagent, New England Biolabs
Supplier: New England Biolabs
Monarch®
T2011L
103529-150EA 124 USD InStock
103529-150
Monarch® DNA/RNA Protection Reagent, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Monarch® DNA/RNA Protection Reagent is a component of the Monarch® Total RNA Miniprep Kit. It is a solution for the preservation of nucleic acids in various tissue types.
  • Preserves and stabilizes RNA and DNA
  • A component of the Monarch® Total RNA Miniprep Kit (103529-148)
It is supplied as a 2X concentrate. This reagent does not need to be removed before RNA extraction with the Monarch® Total RNA Miniprep Kit.
Delivery information: Supplied as a 2X concentrate. Supplied in two bottles, each containing 28 ml.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty