Would you like to log in again?
Monarch® Microfuge Tube EcoRack, New England Biolabs
Supplier: New England Biolabs
Monarch®
T5020S
76326-060EA 39 USD InStock
76326-060
Monarch® Microfuge Tube EcoRack, New England Biolabs
Racks Tube Racks
The Monarch® Microfuge Tube EcoRack is a reversible benchtop tube rack made from plastic recovered during the manufacture of Monarch® Nucleic Acid Purification Columns.
  • Versatile, reversible benchtop tube rack
  • Accommodates 1.5 to 2.0 ml tubes on one side, and 0.5 ml tubes on the reverse side
  • Manufactured using plastic recovered during the manufacture of NEB’s Monarch® Nucleic Acid Purification Columns
Plastic that would otherwise be discarded during the injection molding process is recovered and remolded into this useful lab accessory that can hold up to 48 tubes.
Packaging: The tube racks are sold in packs of 2.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty