Would you like to log in again?
ProLysol Disinfectant Spray, Crisp Linen Scent
Supplier: Essendant
RAC74828CT
76452-782CT 266.55 USD InStock
76452-782
ProLysol Disinfectant Spray, Crisp Linen Scent
Cleaning Agents Disinfectants
This tuberculocidal, virucidal, fungicidal, and bactericidal formula eliminates odors and prevents the growth of mold and mildew.
  • Tuberculocidal, virucidal, fungicidal, bactericidal aerosol spray
  • Eliminates odors and prevents the growth of mold and mildew
  • Meets AOAC Germicidal Spray Product Test standards for hospital aerosol disinfectants
This tuberculocidal, virucidal, fungicidal, and bactericidal formula kills 99.9% of germs on hard, nonporous surfaces and is effective against over 100 microorganisms, including Influenza A Virus, MRSA, Norovirus, Rhinovirus, Poliovirus Type 1, Hepatitis A virus, Hepatitis B virus, and HIV-1 (AIDS Virus) when used as directed. Effective against Mycobacterium bovis BCG (TB) in two minutes (pre-cleaned surfaces). Use on showers, sinks, countertops, around toilet areas, toys, telephones, garbage bins, waiting rooms and in laboratories.
Chemical compound: Alkyl (50% C14, 40% C12, 10% C16) Dimethyl Benzyl Ammonium Saccharinate; Ethanol.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty