Would you like to log in again?
HumaMatrix Native Human-Derived ECM, Lyophilized, Humabiologics
Supplier: Humabiologics
HML-100MG HML-50MG HML-10MG
76452-668EA 600.3 USD InStock
76452-668 76452-666 76452-664
HumaMatrix Native Human-Derived ECM, Lyophilized, Humabiologics
Cell Culture Supplements and Additives
Clinically relevant native human extracellular matrix (ECM) for hydrogel and bioink applications.
  • Native human extracellular matrix (ECM)
  • Clinically relevant and xeno-free for regenerative medicine applications
  • Lyophilized for tailored 3D hydrogel or bioink applications
  • Supports various cell attachments
HumaMatrix is isolated from nutrient-rich human tissue sourced strictly from tissue partners who comply with the requirements for transplantable human tissue. HumaMatrix has undergone a proprietary manufacturing method to preserve native matrix proteins and growth factors. HumaMatrix is supplied lyophlized for custom hydrogel, bioprinting, and cell culture applications.
HumaMatrix addresses the growing need for more clinically relevant and animal-free human tissue constructs and models. Each lot of HumaMatrix has undergone extensive testing to meet high safety and quality requirements for translational research needs.
All tissue has been tested for safety and found negative for communicable diseases and infectious agents.
Ordering information: HumaMatrix is available in 10, 50 and 100 mg.
Caution: HumaMatrix is for research use only and is not intended for human use, diagnosis, screening, household, food or other uses.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty