Would you like to log in again?
VWR® High Performance Undercounter Refrigerator
Supplier: VWR
VWR®
VSU-4RW-N6
76423-860EA 2850.34 USD InStock
76423-860
VWR® High Performance Undercounter Refrigerator
Refrigerators
This Energy Star certified, ADA compliant undercounter refrigerator is designed to meet the current and upcoming CDC, NSF, and ANSI requirements for strict temperature control.
  • Temperature control and ±3 °C uniformity
  • Security and alarm notification
  • Self closing glass door
  • SNAP compliant refrigerant
  • Optional stacking kit
Current and emerging guidelines from the CDC, NSF, and ANSI involve strict storage requirements for vaccines and biologics. The VWR® High Performance undercounter refrigerator is designed to meet these evolving guidelines as they pertain to temperature control, uniformity, and recovery. This unit is UL 471 listed, and meets all electrical standards required by most equivalent Nationally Recognized Testing Laboratories (NRTL).
Strict temperature control keeps stored product within the CDC recommended 2 to 8 °C temperature range for optimal vaccine storage. Many vaccines risk inactivation if allowed to freeze. The undercounter unit is designed to prevent interior temperature from dropping below 2 °C at any location within the cabinet, even if the unit is empty. By contrast, interior temperatures are engineered to cool back to setpoint temperatures within 15 minutes of a 3 minutes door opening.
This Energy Star certified undercounter unit provides energy efficiency and sustainability, without compromising performance. Traditional HFC refrigerants have been replaced with environmentally friendly HC refrigerants to meet Significant New Alternatives Policy (SNAP) guidelines for low global warming and ozone depleting substances.
Certifications: Meets current and emerging CDC, NSF, and ANSI guidelines; ADA compliant; UL 471 listed; SNAP compliant refrigerants.
Ordering information: Supplied without accessories, optional blackout panel and stacking kit are available to order separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty