Would you like to log in again?
Calf Alkaline Phosphatase (from Intestine)
Supplier: Promega
M1821
PAM1821EA 123.93 USD InStock
PAM1821
Calf Alkaline Phosphatase (from Intestine)
Enzymes
Alkaline Phosphatase catalyzes the hydrolysis of 5'-phosphate groups from DNA, RNA and ribo- and deoxyribonucleoside triphosphates.
  • For labeling in vitro and ligation
  • Catalyzes the Hydrolysis of 5´ Phosphate Groups from DNA, RNA, and r- and dNTPs
  • Available at high concentration; Cat.# M2825 contains 1,000 units of CIAP at 20u/microl
  • Qualified for blue/white cloning; our assay provides a higher level of quality control for enzymes used in cloning applications
Alkaline Phosphatase, Calf Intestinal (CIAP), catalyzes the hydrolysis of 5'-phosphate groups from DNA, RNA, and ribo- and deoxyribonucleoside triphosphates. This enzyme is used to prevent recircularization and religation of linearized cloning vector DNA by removing phosphate groups from both 5'-termini and may also be used for the dephosphorylation of 5' phosphorylated ends of DNA or RNA for subsequent labeling with [32P]ATP and T4 Polynucleotide Kinase. CIAP is active on 5' overhangs, 5' recessed and blunt ends.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty