Would you like to log in again?
Whatman™ Glass Microfibre Filters for Personal Air Samplers, Grade GF/A, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
18208296 18208296
76423-726PK 148.78 USD InStock
CA76423-726 76423-726
Whatman™ Glass Microfibre Filters for Personal Air Samplers, Grade GF/A, Whatman products (Cytiva)
Filters Glass Fiber Filters
Grade GF/A card-mounted filters are used in static sample and personal air sampler applications. These aerosol sampling and particulate monitoring filters provide high flow rates and minimal sample interference.
  • Heat resistant up to 500 °C
  • Made of high quality borosilicate glass fibres without added binders
  • Low moisture affinity and high loading capacity
  • Card mounted for use with a continuous air monitor or personal air sampler
Accurate suspended particulate matter analysis is critical in nuclear plants and similar environments.
The GF/A card mounted filter has dimensions of 55×37.5 mm. Please verify that your personal air sampler supports a sample card of this size.
Certifications: Manufactured according to ISO9001:2008 QMS standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty