17-1332-01
17-1332-01
95017-150EA
193.88
USD
InStock
95017-150
CA95017-150L
PlusOne™ Repel-Silane ES, colorless liquid
PlusOne™ Repel-Silane ES
Repel-Silane ES is a 2% solution of dimethyldichlorosilane dissolved in octamethylcyclooctasilane.
- Used in IEF, PAGE and Sequencing
- Soluble in cold water and hot water
It is used to inhibit sticking of polyacrylamide gels, agarose gels, and nucleic acids to glass surfaces.