Would you like to log in again?
Bovine Trypsin
Supplier: G-Biosciences
786-245B 786-245B
78000-052PK 100.59 USD InStock
CA78000-052 78000-052
Bovine Trypsin
Enzymes
Trypsin is a serine endopeptidase that specifically cleaves peptide bonds that are readily identified by mass spectrometry.
  • The resulting trypsin is extremely resistant to autolysis and has a specific activity over 10,000 units/mg protein
  • The maximum activity is in the pH range of 7 to 9 and the activity is reversibly inactivated at pH 4
Native trypsin is prone to autolysis that results in pseudotrypsin, which exhibits a broader proteolytic specificity and trypsin fragments that interfere with sequence analysis.
Unlike other trypsin preparations, our Mass Spectrometry Grade Trypsin is highly stable, and as a result can be stored for a long period without any loss of activity. For convenience, Mass Spectrometry Grade Trypsin (porcine) is supplied lyophilized as 100 or 20µg vials with a specific resuspension buffer.
In addition, the modified trypsin is TPCK treated to inactive the interfering chymotrypsin activity and the resulting protein is affinity purified and lyophilized.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty