Would you like to log in again?
Tyvek® Self-Sealing Stopper Bags, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
Tyvek®
SBMGTVKSS1214
76450-494CS 3055.68 USD InStock
76450-494
Tyvek® Self-Sealing Stopper Bags, Keystone Cleanroom Products
Bags Sterilization Pouches
Self-sealing 1073B Tyvek® / HDPE autoclave bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps. Also a good choice for heavy equipment parts where visibility into the bag is required. Great for applications where a conventional heat sealer is not preferred.
  • Convenient and easy to use self-sealing feature
  • Steam sterilization indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic and lint-free
  • Semi-transparent HDPE film side
  • Consistently strong, double welded factory seals
1073B Tyvek® side permits steam penetration during sterilization. These are a good choice for heavy, or pharmaceutical change parts with sharp corners or where visibility into the bag is required after sterilization. Manufactured and packaged in a controlled cleanroom environment and excellent impact resistance with exceptional microbial barrier.
Certifications: Supplied with Certificates of conformance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty