Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Polypropylene Lab Scoops with Spatula, Eisco
Supplier: Eisco Scientific
CH0631APK10
470328-200EA 13.25 USD InStock
470328-200
Polypropylene Lab Scoops with Spatula, Eisco
Spatulas
Features one scoop end and one spatula end with knife edge. Perfect for a variety of laboratory uses.
  • Made of high quality, chemical and heat resistant polypropylene
  • Autoclavable, reusable
  • Feature one scoop end and one spatula end with knife edge
Blue polypropylene lab scoop with a spoon on one end and flat edge with on the other end. Made of polypropylene which is autoclavable and excellent replacement of metallic scoops. Total Length 150 mm (5.90"), Oval scoop measures 19 mm (0.75") Wide and 26 mm (1.02") long, Flat side measures 20 mm (0.79") wide by 26 mm (1.02") long. Handle is 16 mm (0.63") wide, 3 mm (0.12") thick. These scoops are great for adding small amounts of ingredients and mixing. Pack of 10.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty