Would you like to log in again?
Polypropylene Lab Scoop with Spatula, Eisco
Supplier: Eisco Scientific
CH0631B
470328-202EA 1.85 USD InStock
470328-202
Polypropylene Lab Scoop with Spatula, Eisco
Spatulas
Features one scoop end and one spatula end with knife edge. Perfect for a variety of laboratory uses.
  • Made of high quality, chemical and heat resistant polypropylene
  • Autoclavable, reusable
  • Feature one scoop end and one spatula end with knife edge
Blue polypropylene lab scoop with spoon on one end and flat edge with on the other end. This is made of polypropylene which is autoclavable and excellent replacement of metallic scoops. Total Length: 200 mm (7.87"). Oval scoop measures 21 mm (0.83") wide and 35 mm (1.38") long. Flat side measures 20 mm (0.79") wide by 40 mm (1.57") long. Handle is 17 mm (0.67") wide and 3 mm (0.12") thick.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty