Would you like to log in again?
Dual Ended Stainless Steel Lab Spatula, Eisco
Supplier: EISCO SCIENTIFIC LLC
CH0634C
470328-210EA 2.4 USD InStock
470328-210
Dual Ended Stainless Steel Lab Spatula, Eisco
Spatulas
Features two identical flat spatula ends. Perfect for a variety of laboratory uses.
  • Made of high quality polished stainless steel
  • Autoclavable, reusable
  • Features two identical flat spatula ends
Stainless steel dual ended lab spatula features two identical flat spatula ends, 0.8" wide. Total length is 7.9" Great for adding small amounts of ingredients and mixing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty