Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Dual Ended Teflon Coated Stainless Steel Lab Spatula, Eisco
Supplier: EISCO SCIENTIFIC LLC
CH0634TFC
470328-212EA 4.5 USD InStock
470328-212
Dual Ended Teflon Coated Stainless Steel Lab Spatula, Eisco
Spatulas
Features two identical flat spatula ends. Perfect for a variety of laboratory uses.
  • Made of high quality stainless steel
  • Coated in non-stick Teflon
  • Extremely resistant to chemicals
  • Autoclavable, reusable
  • Feature one scoop end and one spatula end
Teflon coated stainless steel dual ended lab spatula features two identical flat spatula ends, 0.7" wide. Total length is 5.9" Great for adding small amounts of ingredients and mixing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty