Would you like to log in again?
Stainless Steel Lab Scoop with Spatula, Eisco
Supplier: Eisco Scientific
CH0635C
470328-214EA 3.85 USD InStock
470328-214
Stainless Steel Lab Scoop with Spatula, Eisco
Spatulas
Features one scoop end and one flat spatula end. Perfect for a variety of laboratory uses.
  • Made of high quality polished stainless steel
  • Autoclavable, reusable
  • Feature one scoop end and one spatula end
Stainless steel lab scoop with spoon on one end and flat edge with on the other end. Total legnth is 9", Spatula measures 0.3" wide, scoop measures 0.5" wide. These scoops are great for adding small amounts of ingredients and mixing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty